Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubc-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061186.5 | NP_493587.1 | 2 - 3 | 232 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |