Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026695.4 | NP_001021866.1 | 8 - 9 | 12988 | 100 | |
NM_061199.6 | NP_493600.2 | 7 - 8 | 12851 | 100 | ||
NM_061200.6 | NP_493601.2 | 7 - 8 | 12846 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |