Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061219.7 | NP_493620.2 | 7 - 8 | 1203 | 71 |
Gene Symbol: F33H2.8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061220.6 | NP_493621.2 | 14 - 15 | 2695 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |