Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cwn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061267.5 | NP_493668.1 | 1 - 2 | 177 | 94 | |
NR_132572.1 | 1 - 2 | 156 | 94 | |||
GenBank mRNA | X72941.1 | - | 171 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |