Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cat-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306554.2 | NP_001293483.1 | 1 - 2 | 48 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |