Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313598.3 | NP_001300527.1 | 1 - 2 | 107 | 98 | |
NM_061312.5 | NP_493713.2 | 4 - 5 | 567 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |