Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prp-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061345.10 | NP_493746.1 | 8 - 9 | 1797 | 75 | |
GenBank mRNA | U09415.1 | - | 8 - 9 | 1808 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |