Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdz-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061380.7 | NP_493781.1 | 4 - 5 | 562 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |