Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: somi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373144.3 | NP_001360520.1 | 4 - 5 | 1613 | 77 | |
NM_001373145.2 | NP_001360558.1 | 3 - 4 | 1516 | 77 | ||
NM_001383439.1 | 5 - 6 | 1712 | 77 | |||
NM_073919.6 | NP_506320.3 | 4 - 5 | 1602 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |