Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxc-40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318282.3 | NP_001305211.1 | 2 - 3 | 901 | 76 | |
NM_001383782.1 | 4 - 5 | 1295 | 76 | |||
NM_061474.7 | NP_493875.1 | 4 - 5 | 1222 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |