Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sut-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129167.3 | NP_001122639.1 | 3 - 4 | 194 | 68 | |
NM_061516.5 | NP_493917.2 | 3 - 4 | 188 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |