Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pod-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267098.3 | NP_001254027.1 | 9 - 10 | 6259 | 63 | |
NM_001381323.1 | NP_001368018.1 | 10 - 11 | 6397 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |