Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027006.6 | NP_001022177.1 | 3 - 4 | 575 | 59 | |
NM_001027007.3 | NP_001022178.1 | 3 - 4 | 575 | 59 | ||
NM_001322704.3 | NP_001309559.1 | 4 - 5 | 726 | 59 | ||
NM_001322707.3 | NP_001309561.1 | 4 - 5 | 726 | 59 | ||
GenBank mRNA | AF183400.1 | - | 3 - 4 | 575 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |