Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alh-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027041.8 | NP_001022212.1 | 6 - 7 | 1486 | 84 | |
NM_001047317.6 | NP_001040782.1 | 3 - 4 | 1055 | 84 | ||
NM_061545.7 | NP_493946.1 | 6 - 7 | 1478 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |