Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jip-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038242.6 | NP_001033331.1 | 9 - 10 | 1393 | 106 | |
NM_001038243.6 | NP_001033332.1 | 7 - 8 | 931 | 106 | ||
NM_001377691.1 | NP_001364697.1 | 17 - 18 | 2836 | 106 | ||
NM_001377692.2 | NP_001364548.1 | 17 - 18 | 2827 | 106 | ||
NM_001377693.2 | NP_001364549.1 | 9 - 10 | 1408 | 106 | ||
NM_001377694.2 | NP_001364547.1 | 7 - 8 | 946 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |