Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cogc-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038252.4 | NP_001033341.1 | 1 - 2 | 432 | 78 | |
NM_061565.6 | NP_493966.1 | 2 - 3 | 767 | 78 | ||
NM_061566.7 | NP_493967.1 | 1 - 2 | 432 | 78 |
Gene Symbol: Y51H7C.41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381330.2 | NP_001368658.1 | 4 - 5 | 1315 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |