Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: math-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061600.5 | NP_494001.3 | 2 - 3 | 512 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |