Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bath-46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061603.3 | NP_494004.2 | 4 - 5 | 692 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |