Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bath-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061604.6 | NP_494005.2 | 4 - 5 | 804 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |