Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bath-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061606.5 | NP_494007.1 | 2 - 3 | 529 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |