Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btb-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061627.3 | NP_494028.2 | 2 - 3 | 376 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |