Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061633.1 | NP_494034.1 | 2 - 3 | 594 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |