Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F36H5.18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377701.1 | NP_001364538.1 | 1 - 2 | 397 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |