Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: math-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061711.3 | NP_494112.1 | 2 - 3 | 394 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |