Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vab-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061875.3 | NP_494276.2 | 17 - 18 | 3020 | 154 | |
GenBank mRNA | AY273823.1 | - | 17 - 18 | 3007 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |