Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129141.3 | NP_001122613.1 | 2 - 3 | 663 | 60 | |
NM_061876.8 | NP_494277.3 | 7 - 8 | 1236 | 60 | ||
NM_061877.7 | NP_494278.3 | 7 - 8 | 1241 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |