Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381350.1 | NP_001368720.1 | 11 - 12 | 2160 | 89 | |
NM_001383801.1 | 13 - 14 | 2436 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |