Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sup-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061932.5 | NP_494333.1 | 3 - 4 | 444 | 73 | |
GenBank mRNA | AF083652.1 | - | 3 - 4 | 388 | 73 | |
AY357729.1 | - | 3 - 4 | 458 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |