Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061971.5 | NP_494372.2 | 1 - 2 | 158 | 122 | |
GenBank mRNA | AJ505020.1 | - | 1 - 2 | 99 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |