Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377741.1 | NP_001364794.1 | 2 - 3 | 700 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |