Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001361852.3 | NP_001348701.1 | 10 | 1243 | 136 | |
NM_001361853.3 | NP_001348700.1 | 10 - 11 | 1313 | 136 | ||
NM_001361854.3 | NP_001348702.1 | 11 | 1276 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Amplicon spans exons and probe does not span exons |