Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scl-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062095.6 | NP_494496.1 | 2 - 3 | 522 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |