Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apd-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062169.6 | NP_494570.1 | 11 - 12 | 2924 | 128 | |
NM_062170.5 | NP_494571.1 | 7 - 8 | 2204 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |