Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062208.3 | NP_494609.1 | 7 - 8 | 717 | 62 |
Gene Symbol: srx-98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377766.2 | NP_001364762.1 | 7 - 8 | 725 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |