Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062226.1 | NP_494627.1 | 3 - 4 | 987 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |