Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062277.4 | NP_494678.3 | 11 - 12 | 1841 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |