Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhgf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381382.1 | NP_001368191.1 | 2 - 3 | 519 | 61 | |
NM_001381384.1 | NP_001367992.1 | 2 - 3 | 250 | 61 | ||
NM_001383815.1 | 2 - 3 | 253 | 61 | |||
GenBank mRNA | JQ763434.1 | - | 1 - 2 | 93 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |