Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhgf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381382.1 | NP_001368191.1 | 9 - 10 | 1738 | 82 | |
NM_001381384.1 | NP_001367992.1 | 9 - 10 | 1469 | 82 | ||
NM_001383815.1 | 9 - 10 | 1472 | 82 | |||
GenBank mRNA | JQ763434.1 | - | 8 - 9 | 1312 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |