Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srm-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074154.5 | NP_506555.2 | 2 - 3 | 496 | 127 | |
NM_182385.4 | NP_872185.2 | 2 - 3 | 402 | 127 |
Gene Symbol: srm-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028771.4 | NP_001023942.1 | 7 - 8 | 1843 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |