Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nlg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313376.3 | NP_001300305.1 | 14 - 15 | 2353 | 96 | |
NM_001313406.3 | NP_001300335.1 | 7 - 8 | 1225 | 96 | ||
NM_182454.4 | NP_872254.1 | 14 - 15 | 2415 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |