Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001364060.1 | NP_001350986.1 | 13 - 14 | 1668 | 113 | |
NM_001364061.1 | NP_001350985.1 | 13 - 14 | 1668 | 113 | ||
NM_001364062.1 | NP_001350987.1 | 13 - 14 | 1668 | 113 | ||
NM_001364063.2 | NP_001350984.1 | 13 - 14 | 1668 | 113 | ||
NM_001364064.3 | NP_001350988.1 | 13 - 14 | 1668 | 113 | ||
NM_062385.7 | NP_494786.5 | 13 - 14 | 1668 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |