Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sre-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062431.2 | NP_494832.2 | 1 - 2 | 291 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |