Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ulp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318311.3 | NP_001305240.1 | 10 | 1139 | 69 | |
NM_062513.5 | NP_494914.1 | 12 - 13 | 1685 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |