Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313625.4 | NP_001300554.1 | 1 - 2 | 256 | 76 | |
NM_001383826.1 | 2 - 3 | 455 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |