Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pld-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062538.7 | NP_494939.1 | 10 - 11 | 3219 | 137 | |
GenBank mRNA | AB028889.1 | - | 10 - 11 | 3241 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |