Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pld-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062538.7 | NP_494939.1 | 17 - 18 | 4240 | 66 | |
GenBank mRNA | AB028889.1 | - | 17 - 18 | 4262 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |