Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026742.5 | NP_001021913.1 | 6 - 7 | 1053 | 69 | |
NM_062545.8 | NP_494946.2 | 6 - 7 | 1032 | 69 | ||
NM_062546.8 | NP_494947.2 | 6 - 7 | 1021 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |