Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srb-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062555.4 | NP_494956.1 | 7 - 8 | 962 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |