Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srb-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368586.3 | NP_001355415.1 | 2 - 3 | 449 | 111 | |
NM_062556.3 | NP_494957.1 | 2 - 3 | 450 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |