Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dsh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313591.3 | NP_001300520.1 | 9 - 10 | 1392 | 83 | |
NM_001313592.3 | NP_001300521.1 | 9 - 10 | 2817 | 83 | ||
NM_001383828.1 | 13 - 14 | 2715 | 83 | |||
NM_062577.8 | NP_494978.4 | 14 - 15 | 2091 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |